missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)aniline, 98%

Catalog No. p-7044165
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL0495006 5 g
AAL0495014 25 g
2 options

Catalog No. AAL0495006

Supplier: Thermo Scientific Chemicals L0495006

Only null left

CAS: 104-96-1 | C7H9NS | 139.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-96-1
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.22
MDL Number MFCD00007889
InChI Key YKFROQCFVXOUPW-UHFFFAOYSA-N
Synonym 4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine
PubChem CID 66038
IUPAC Name 4-methylsulfanylaniline
SMILES CSC1=CC=C(N)C=C1

Specifications

Density 1.12
Boiling Point 272°C to 273°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.639
Quantity 5 g
UN Number UN2810
Beilstein 774506
Sensitivity Light sensitive
Formula Weight 139.23
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)aniline
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-256-3
RTECSNumber BY6302000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only