accessibility menu, dialog, popup

UserName

CAS RN 104-96-1

IUPAC Name: 4-(methylsulfanyl)aniline
Synonyms: benzenamine, 4-(methylthio)-
Molecular Weight (g/mol): 139.22
Molecular Formula: C7H9NS
InChi Key: YKFROQCFVXOUPW-UHFFFAOYSA-N
SMILES: CSC1=CC=C(N)C=C1
Boiling Point 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)