missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-sulfonyl chloride, 95%, Thermo Scientific™

Catalog No. p-7032321
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33732MD 250mg
AAH3373203 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33732MD Supplier Thermo Scientific Chemicals Supplier No. H33732MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 953070-51-4
Molecular Formula C4H4ClNO2S2
Molecular Weight (g/mol) 197.651
MDL Number MFCD09864968
InChI Key AWTJUYXVSWZCIK-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl, chloro 4-methyl 1,3-thiazol-5-yl sulfone
PubChem CID 18537519
IUPAC Name 4-methyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC=N1)S(=O)(=O)Cl

Specifications

Quantity 250mg
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 197.66
Percent Purity 95%
Chemical Name or Material 4-Methylthiazole-5-sulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only