missing translation for 'onlineSavingsMsg'
Learn More

4-Methylthiazole-5-carboxaldehyde, 97%

Catalog No. p-7041293
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6161303 1 g
AAH6161306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6161303 Supplier Thermo Scientific Chemicals Supplier No. H6161303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 82294-70-0 | C5H5NOS | 127.161 g/mol

4-Methylthiazole-5-carboxaldehyde is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylthiazole-5-carboxaldehyde is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 82294-70-0
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD07368277
InChI Key JJVIEMFQPALZOZ-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carbaldehyde, 4-methyl-5-thiazolecarboxaldehyde, 4-methylthiazole-5-carboxaldehyde, 4-methyl-5-formylthiazole, 4-methyl-5-thiazolecarbaldehyde, 5-formyl-4-methylthiazole, 4-methylthiazole-5-aldehyde, 4-methyl-5-thiazolyl aldehyde, 5-thiazolecarboxaldehyde, 4-methyl, 4-methyl-5-thiazole carboxaldehyde
PubChem CID 581339
IUPAC Name 4-methyl-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC=N1)C=O

Specifications

Melting Point 74°C to 78°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 127.16
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxaldehyde
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only