missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%

Catalog No. AC424960025
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424960025 2.5 g Glass bottle
AC424960100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC424960025 Supplier Thermo Scientific Chemicals Supplier No. 424960025

CAS: 17852-67-4 | C7H10N2O2S·HCl | 222.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17852-67-4
Molecular Formula C7H10N2O2S·HCl
Molecular Weight (g/mol) 222.69
MDL Number MFCD00216494
InChI Key QVCMFSJTNVQJFG-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenyl hydrazine hydrochloride, 4-methylsulfonyl phenylhydrazine hydrochloride, 4-methylsulphonyl phenylhydrazine hydrochloride, 4-methylsulphonylphenylhydrazine hydrochloride, 4-methanesulfonylphenyl hydrazine hydrochloride, 4-methylsulfonylphenylhydrazine hydrochloride, hydrazine, 4-methylsulfonyl phenyl-, monohydrochloride, 4-methylsulfonyl phenylhydrazine, chloride, 4-methylsulfonyl phenylhydrazine hydrochloric acid
PubChem CID 2735181
IUPAC Name (4-methylsulfonylphenyl)hydrazine;hydrochloride
SMILES CS(=O)(=O)C1=CC=C(C=C1)NN.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 202°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NHNC6H4SO2CH3·HCl
Quantity 2.5 g
Solubility Information Solubility in water: soluble
Formula Weight 222.69
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)phenylhydrazine hydrochloride
Show More Show Less

RUO – Research Use Only