missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenylacetic acid, 97%

Catalog No. AAL1950406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1950406 5 g
AAL1950414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1950406 Supplier Thermo Scientific Chemicals Supplier No. L1950406
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90536-66-6 | C9H10O4S | 214.235 g/mol

4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 90536-66-6
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00216495
InChI Key HGGWOSYNRVOQJH-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenylacetic acid, 4-methylsulphonylphenylacetic acid, 2-4-methylsulfonyl phenyl acetic acid, 4-mesylphenylacetic acid, 4-methanesulfonylphenyl acetic acid, 4-methylsulfonyl phenyl acetic acid, 4-methanesulfonyl phenylacetic acid, 2-4-methanesulfonylphenyl acetic acid, 4-methylsulfonylphenylacetic acid
PubChem CID 572345
IUPAC Name 2-(4-methylsulfonylphenyl)acetic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 134°C to 138°C
Quantity 5 g
Beilstein 2646649
Solubility Information Slightly soluble in water.
Formula Weight 214.24
Percent Purity 97%
Chemical Name or Material 4-(Methylsulfonyl)phenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only