missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Methylsulfonyl)benzyl Bromide 97.0+%, TCI America™
Supplier: TCI America M24745G
Specifications
| 4-(Methylsulfonyl)benzyl Bromide | |
| 97°C | |
| C8H9BrO2S | |
| 5 g | |
| 1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix | |
| CS(=O)(=O)C1=CC=C(C=C1)CBr | |
| 249.122 | |
| 249.12 | |
| Crystalline Powder |
| 53606-06-7 | |
| White-Yellow | |
| MFCD00185849 | |
| 3261 | |
| HGKPAXHJTMHWAH-UHFFFAOYSA-N | |
| 1-(bromomethyl)-4-methylsulfonylbenzene | |
| 2733581 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 53606-06-7 | |
| 249.122 | |
| HGKPAXHJTMHWAH-UHFFFAOYSA-N | |
| 2733581 | |
| CS(=O)(=O)C1=CC=C(C=C1)CBr |
| C8H9BrO2S | |
| MFCD00185849 | |
| 1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix | |
| 1-(bromomethyl)-4-methylsulfonylbenzene |
Safety and Handling
TSCA : No