accessibility menu, dialog, popup

UserName

CH3SOOCl

CAS RN 53606-06-7

IUPAC Name: 1-(chloromethyl)-4-methanesulfonylbenzene
Synonyms: 1-(chloromethyl)-4-methanesulfonylbenzene
Molecular Weight (g/mol): 204.67
Molecular Formula: C8H9ClO2S
InChi Key: LXPRVXKHIXWBJZ-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(CCl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)