missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzyl Bromide 97.0+%, TCI America™

Catalog No. p-7133240
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24741G 1 g
M24745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24741G Supplier TCI America Supplier No. M24741G
Only null left

Chemical Identifiers

CAS 53606-06-7
Molecular Formula C8H9BrO2S
Molecular Weight (g/mol) 249.122
MDL Number MFCD00185849
InChI Key HGKPAXHJTMHWAH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix
PubChem CID 2733581
IUPAC Name 1-(bromomethyl)-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)CBr

Specifications

Melting Point 97°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 249.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)benzyl Bromide