missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzoic Acid 98.0+%, TCI America™

Catalog No. M19325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19325G 5 g
M193225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19325G Supplier TCI America Supplier No. M19325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4052-30-6
Molecular Formula C8H8O4S
Molecular Weight (g/mol) 200.208
MDL Number MFCD00007564
InChI Key AJBWNNKDUMXZLM-UHFFFAOYSA-N
Synonym 4-methylsulfonyl benzoic acid, p-methylsulfonylbenzoic acid, 4-methanesulfonyl-benzoic acid, 4-methylsulphonylbenzoic acid, 4-methylsulphonyl benzoic acid, 4-methanesulfonylbenzoic acid, benzoic acid, 4-methylsulfonyl, p-methylsulphonyl benzoic acid, 4-methanesulfonyl benzoic acid
PubChem CID 77670
IUPAC Name 4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 265°C
Color White
Quantity 5 g
Formula Weight 200.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)benzoic Acid