missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulfamoylbenzeneboronic acid, 97%

Catalog No. AAH52493MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52493MD 250 mg
AAH5249303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52493MD Supplier Thermo Scientific Chemicals Supplier No. H52493MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 226396-31-2 | C7H10BNO4S | 215.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 226396-31-2
Molecular Formula C7H10BNO4S
Molecular Weight (g/mol) 215.03
MDL Number MFCD06659864
InChI Key DOQOQZHSIBBHMY-UHFFFAOYSA-N
Synonym 4-n-methylsulfamoyl phenyl boronic acid, methyl 4-boronobenzenesulfonamide, 4-methylsulfamoyl phenyl boronic acid, 4-methylaminosulphonyl benzene boronic acid, 4-n-methylsulfamoyl phenylboronic acid, 4-methylsulfamoyl phenylboronic acid, 4-methylsulfamoylphenylboronic acid, boronic acid, 4-methylamino sulfonyl phenyl, acmc-209fxs
PubChem CID 44118763
IUPAC Name [4-(methylsulfamoyl)phenyl]boronic acid
SMILES CNS(=O)(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 136°C to 140°C
Quantity 250 mg
Formula Weight 215.03
Percent Purity 97%
Chemical Name or Material 4-Methylsulfamoylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only