missing translation for 'onlineSavingsMsg'
Learn More

4-Méthylpipéridine 98,0+%, TCI America™

Numéro de catalogue. P044525ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
P044525ML 25 mL
P0445500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. P044525ML Fournisseur TCI America Code fournisseur P044525ML

Identifiants chimiques

CAS 626-58-4
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00006005
InChI Key UZOFELREXGAFOI-UHFFFAOYSA-N
Synonym 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline
PubChem CID 69381
IUPAC Name 4-méthylpipéridine
SMILES CC1CCNCC1

Spécifications

Color Jaune
Boiling Point 127°C
Quantity 25 mL
UN Number 1993
Formula Weight 99.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquide
Chemical Name or Material 4-Methylpiperidine
EINECSNumber (5)-0768
TSCA Yes