missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4-Methylpiperidine solution

Numéro de catalogue. 7921524L
Encompass
Modifier l'affichage
Click to view available options
Quantité:
2 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
7921524L 4 L
7921522L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. 7921524L Fournisseur Sigma Aldrich Code fournisseur 7921524L
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methylpiperidine solution (20 % in dmf) can be used:
  • In the deprotection of f-moc protecting group which is attached to free amine
  • In the determination of resin-bound amino acid by UV-fmoc-quantitation

Spécifications

CAS 626-58-4
Densité 0.920 g/mL (at 25°C)
Formule moléculaire C6H13N
Indice de réfraction n20/D 1.433
Formule linéaire C6H13N
Numéro MDL MFCD00006005
Quantité 4 L
Synonyme 4-Methylpiperidine solution
Poids moléculaire (g/mol) 99.17