missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4-Methylmorpholine

Numéro de catalogue. 4077041L
Encompass
Modifier l'affichage
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
4077041L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 4077041L Fournisseur Sigma Aldrich Code fournisseur 4077041L
Il en reste null

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Methylmorpholine

Spécifications

CAS 109-02-4
Melting Point -66°C (lit.)
Density 0.92 g/mL (at 25°C (literature))
Boiling Point 115°C to 116°C (750 mmHg)
Molecular Formula C5H11NO
Refractive Index n20/D 1.435 (literature)
Linear Formula C5H11NO
MDL Number MFCD00006175
Quantity 1 L
Synonym NMM
Molecular Weight (g/mol) 101.15
Percent Purity ≥99.5%
Afficher plus de résultats Afficher moins de résultats