missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylcyclohexylamine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 223°C |
| Quantity | 25 g |
| Formula Weight | 149.66 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 4-Methylcyclohexylamine Hydrochloride |
Safety and Handling
| TSCA | Yes |