missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcyclohexanone 98.0+%, TCI America™

Numéro de catalogue. p-7132050
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M019925ML 25 mL
M0199500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M019925ML Fournisseur TCI America Code fournisseur M019925ML
Il en reste null

Identifiants chimiques

CAS 589-92-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00001643
InChI Key VGVHNLRUAMRIEW-UHFFFAOYSA-N
Synonym 4-methylcyclohexanone, cyclohexanone, 4-methyl, 4-methyl-1-cyclohexanone, methyl-4 cyclohexanone-1, p-methyl cyclohexanone, methycyclohexanone, metylocykloheksanon, unii-0l1r78k79s, 4-methyl cyclohexanone, methyl-4 cyclohexanone-1 french
PubChem CID 11525
IUPAC Name 4-methylcyclohexan-1-one
SMILES CC1CCC(=O)CC1

Spécifications

Color Yellow
Boiling Point 171°C
Quantity 25 mL
UN Number 2297
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylcyclohexanone
EINECSNumber (3)-2411
RTECSNumber GW1750200
TSCA Yes