missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 4-Methylcyclohexanol, mixture of cis and trans

Numéro de catalogue. 1530951L
Encompass
Modifier l'affichage
Click to view available options
Quantité:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
1530951L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 1530951L Fournisseur Sigma Aldrich Code fournisseur 1530951L

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Methylcyclohexanol, mixture of cis and trans was used to study the effects of 4-Methylcyclohexanol on oviposition.

Spécifications

CAS 589-91-3
Densité 0.914 g/mL (at 25°C (literature))
Point d’ébullition 171°C to 173°C (lit.)
Formule moléculaire C7H14O
Indice de réfraction n20/D 1.458 (literature)
Formule linéaire CH3C6H10OH
Numéro MDL MFCD00001449
Quantité 1 L
Synonyme Hexahydro-p-cresol
Poids moléculaire (g/mol) 114.19
Pourcentage de pureté 98%
Afficher plus de résultats Afficher moins de résultats