missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbiphenyl, 98%

Catalog No. p-7022093
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1470206 5 g
AAA1470214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1470206 Supplier Thermo Scientific Chemicals Supplier No. A1470206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 644-08-6 | C13H12 | 168.24 g/mol

4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbiphenyl, is used as a flavoring ingredient in food industry. It is also used as an intermediate in agrochemcial, dyestuff, flavor and fragrance, pharmaceutical, syntheses material. It is also used as an OLED intermediate.

Solubility
Insoluble in water. Soluble in alcohol and ether, very soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 644-08-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.24
MDL Number MFCD00008544
InChI Key ZZLCFHIKESPLTH-UHFFFAOYSA-N
Synonym 4-methylbiphenyl, 4-phenyltoluene, 4-methyl-1,1'-biphenyl, p-methylbiphenyl, p-methyldiphenyl, 4-methyldiphenyl, 1,1'-biphenyl, 4-methyl, biphenyl, 4-methyl, p-phenyltoluene, 4-methylphenyl benzene
PubChem CID 12566
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 44°C to 48°C
Density 1.015
Boiling Point 267°C to 268°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1904468
Solubility Information Insoluble in water. Soluble in alcohol and ether,very soluble in organic solvents.
IUPAC Name 1-methyl-4-phenylbenzene
Formula Weight 168.24
Percent Purity 98%
Chemical Name or Material 4-Methylbiphenyl
Show More Show Less
EINECSNumber 211-409-0
RTECSNumber DV5460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only