missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl Phenyl Ketone 97.0+%, TCI America™

Catalog No. M16395G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16395G 5 g
M163925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16395G Supplier TCI America Supplier No. M16395G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2430-99-1
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
InChI Key WILFDKCJCDVGQX-UHFFFAOYSA-N
Synonym 2-(p-Tolyl)acetophenone, p-Tolylmethyl Phenyl Ketone
PubChem CID 242473
IUPAC Name 2-(4-methylphenyl)-1-phenylethanone
SMILES CC1=CC=C(C=C1)CC(=O)C2=CC=CC=C2

Specifications

Melting Point 95°C
Color White
Boiling Point 155°C
Quantity 5 g
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzyl Phenyl Ketone