missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylbenzyl Alcohol 99.0+%, TCI America™
Supplier: TCI America M0162100G
Specifications
| 4-Methylbenzyl Alcohol | |
| 60°C | |
| 116°C | |
| MFCD00004664 | |
| 4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol | |
| CC1=CC=C(C=C1)CO | |
| 122.167 | |
| CHEBI:1895 | |
| ≥99.0% (GC) |
| 589-18-4 | |
| White-Yellow | |
| C8H10O | |
| 100 g | |
| KMTDMTZBNYGUNX-UHFFFAOYSA-N | |
| (4-methylphenyl)methanol | |
| 11505 | |
| 122.17 | |
| Crystalline Powder |
Chemical Identifiers
| 589-18-4 | |
| 122.167 | |
| KMTDMTZBNYGUNX-UHFFFAOYSA-N | |
| 11505 | |
| (4-methylphenyl)methanol |
| C8H10O | |
| MFCD00004664 | |
| 4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol | |
| CHEBI:1895 | |
| CC1=CC=C(C=C1)CO |
Safety and Handling
EINECSNumber : (3)-1049
RTECSNumber : DO9370000
TSCA : Yes