missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl Alcohol 99.0+%, TCI America™

Catalog No. p-7132027
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M016225G 25 g
M0162100G 100 g
M0162500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M016225G Supplier TCI America Supplier No. M016225G
Only null left

Chemical Identifiers

CAS 589-18-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004664
InChI Key KMTDMTZBNYGUNX-UHFFFAOYSA-N
Synonym 4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
PubChem CID 11505
ChEBI CHEBI:1895
IUPAC Name (4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)CO

Specifications

Melting Point 60°C
Color White-Yellow
Boiling Point 116°C
Quantity 25 g
Formula Weight 122.17
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzyl Alcohol
EINECSNumber (3)-1049
RTECSNumber DO9370000
TSCA Yes