accessibility menu, dialog, popup

UserName

CAS RN 589-18-4

IUPAC Name: (4-methylphenyl)methanol
Synonyms: 4-methylbenzyl alcohol P-methyl benzyl alcohol p-methylbenzylalkohol
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: KMTDMTZBNYGUNX-UHFFFAOYSA-N
SMILES: CC1=CC=C(CO)C=C1
Boiling Point 
  • (3)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)