missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl alcohol, 99%

CAS: 589-18-4 | C8H10O | 122.167 g/mol

Supplier:  Thermo Scientific Chemicals A1531522

 View more versions of this product

Catalog No. AAA1531522


Only null left
Explore available promotions

Description

Description

4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.

Solubility
Soluble in methanol, ether and ethanol. Slightly soluble in water.

Notes
Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications

Specifications

4-Methylbenzyl alcohol
57°C to 61°C
217°C
MFCD00004664
1856550
Soluble in methanol,ether and ethanol. Slightly soluble in water.
CC1=CC=C(C=C1)CO
122.167
CHEBI:1895
99%
589-18-4
0.978
C8H10O
100 g
4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
KMTDMTZBNYGUNX-UHFFFAOYSA-N
(4-methylphenyl)methanol
11505
122.17

Chemical Identifiers

589-18-4
122.167
KMTDMTZBNYGUNX-UHFFFAOYSA-N
11505
(4-methylphenyl)methanol
C8H10O
MFCD00004664
4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
CHEBI:1895
CC1=CC=C(C=C1)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-639-1

RTECSNumber : DO9370000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only