missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl alcohol, 99%

Catalog No. p-7022499
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1531514 25 g
AAA1531522 100 g
AAA1531536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1531514 Supplier Thermo Scientific Chemicals Supplier No. A1531514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 589-18-4 | C8H10O | 122.167 g/mol

4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzyl alcohol is used in fragrances, flavoring and as a laboratory reagent. It is also used as a raw material for the preparation of polycarbonates and 4-methyl-benzaldehyde.

Solubility
Soluble in methanol, ether and ethanol. Slightly soluble in water.

Notes
Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 589-18-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00004664
InChI Key KMTDMTZBNYGUNX-UHFFFAOYSA-N
Synonym 4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
PubChem CID 11505
ChEBI CHEBI:1895
IUPAC Name (4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)CO

Specifications

Melting Point 57°C to 61°C
Density 0.978
Boiling Point 217°C
Quantity 25 g
Beilstein 1856550
Solubility Information Soluble in methanol,ether and ethanol. Slightly soluble in water.
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material 4-Methylbenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-639-1
RTECSNumber DO9370000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only