missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzophenone 95.0+%, TCI America™

Catalog No. M124825G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M124825G 25 g
M1248250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M124825G Supplier TCI America Supplier No. M124825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134-84-9
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008553
InChI Key WXPWZZHELZEVPO-UHFFFAOYSA-N
Synonym 4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
PubChem CID 8652
IUPAC Name (4-methylphenyl)-phenylmethanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 56°C
Color White-Yellow
Boiling Point 328°C
Quantity 25 g
Formula Weight 196.25
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylbenzophenone
EINECSNumber (9)-2127
RTECSNumber DJ1750000
TSCA Yes