missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzoic anhydride, 97%

Catalog No. AAH3395103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3395103 1 g
AAH3395106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3395103 Supplier Thermo Scientific Chemicals Supplier No. H3395103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13222-85-0 | C16H14O3 | 254.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13222-85-0
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.285
MDL Number MFCD00048100
InChI Key BJMLLSSSTGHJJE-UHFFFAOYSA-N
Synonym 4-methylbenzoic anhydride, 4-methylphenyl anhydride, 4-methylbenzoic acid anhydride, p-toluic anhydride, 4-methylbenzene-1-carboxylic anhydride, 4-methylbenzoyl 4-methylbenzoate, bis 4-methylbenzoic anhydride, p-toluylsaureanhydrid, para-toluic anhydride, 4-methylbenzoicanhydride
PubChem CID 139413
IUPAC Name (4-methylbenzoyl) 4-methylbenzoate
SMILES CC1=CC=C(C=C1)C(=O)OC(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 86°C to 89°C
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 254.29
Percent Purity 97%
Chemical Name or Material 4-Methylbenzoic anhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only