missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylaminoantipyrine hydrochloride, ≥95.0% (HPLC); 95.0-105.0% (AT), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 856307-27-2 |
| Linear Formula | C12H15N3 O · HCl |
| MDL Number | MFCD16620361 |
| Synonym | 1,2-Dihydro-1,5-dimethyl-4-(methylamino)-2-phenyl-3 H-pyrazol-3-one monohydrochloride; N-Methylaminophenazone; Noramidopyrine hydrochloride; Noraminopyrine hydrochloride |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 253.73 |
| Percent Purity | ≥95.0% (HPLC); 95.0 to 105.0% (AT) |
| Grade | Analytical Standard |