missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, 95%

Catalog No. AC126180050
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC126180050 5 g Glass bottle
AC126182500 250 g Glass bottle
AC126180010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC126180050 Supplier Thermo Scientific Chemicals Supplier No. 126180050

CAS: 122-00-9 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -19.0°C
Color Colorless to Yellow
Density 1.0050g/mL
Boiling Point 226.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5320 to 1.5350
Linear Formula CH3C6H4COCH3
Quantity 5 g
Beilstein 07, 307
Specific Gravity 1.005
Solubility Information Solubility in water: 0.37g/L (15°C)
Formula Weight 134.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4'-Methylacetophenone, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 204-514-8
RTECSNumber AM9463000
TSCA TSCA

RUO – Research Use Only