missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl(thiobenzamide), 97%

Catalog No. AAB2032003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2032003 1 g
AAB2032006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2032003 Supplier Thermo Scientific Chemicals Supplier No. B2032003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2362-62-1 | C8H9NS | 151.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2362-62-1
Molecular Formula C8H9NS
Molecular Weight (g/mol) 151.227
MDL Number MFCD00173750
InChI Key QXYZSNGZMDVLKN-UHFFFAOYSA-N
Synonym 4-methylbenzothioamide, 4-methyl thiobenzamide, 4-methylthiobenzamide, 4-methylbenzene-1-carbothioamide, 4-methyl-thiobenzamide, 4-methylbenzenethioamide, benzenecarbothioamide, 4-methyl, amino 4-methylphenyl methane-1-thione, 4-pmtb, p-methylthiobenzoylamide
PubChem CID 737209
IUPAC Name 4-methylbenzenecarbothioamide
SMILES CC1=CC=C(C=C1)C(=S)N

Specifications

Melting Point 166°C to 169°C
Quantity 1 g
Formula Weight 151.23
Percent Purity 97%
Chemical Name or Material 4-Methyl(thiobenzamide)
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only