missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-7-morpholino-8-azacoumarin 98.0+%, TCI America™

Catalog No. M2937200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2937200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2937200MG Supplier TCI America Supplier No. M2937200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 57980-07-1
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.266
MDL Number MFCD18803635
InChI Key UGHFUCYBDDPFJX-UHFFFAOYSA-N
Synonym 4-Methyl-7-morpholino-2H-pyrano[2,3-b]pyridin-2-one
PubChem CID 93904
IUPAC Name 4-methyl-7-morpholin-4-ylpyrano[2,3-b]pyridin-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=N2)N3CCOCC3

Specifications

Melting Point 189°C
Quantity 200 mg
Formula Weight 246.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-7-morpholino-8-azacoumarin