missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-vinylthiazole, 98+%

Catalog No. AAB2171906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2171906 5 g
AAB2171914 25 g
AAB2171922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2171906 Supplier Thermo Scientific Chemicals Supplier No. B2171906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1759-28-0 | C6H7NS | 125.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1759-28-0
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00005337
InChI Key QUAMMXIRDIIGDJ-UHFFFAOYSA-N
Synonym 4-methyl-5-vinylthiazole, thiazole, 5-ethenyl-4-methyl, 4-methyl-5-vinyl thiazole, 5-ethenyl-4-methylthiazole, thiazole, 4-methyl-5-vinyl, vinylsulfurol, vinyl sulfurol, fema no. 3313, 5-vinyl-4-methylthiazole, 4-methyl-5-vinyl-thiazole
PubChem CID 15654
IUPAC Name 5-ethenyl-4-methyl-1,3-thiazole
SMILES CC1=C(SC=N1)C=C

Specifications

Melting Point -15°C
Density 1.095
Boiling Point 67°C to 68°C (11 mmHg)
Flash Point 70°C (158°F)
Odor Pleasant
Refractive Index 1.568
Quantity 5 g
Beilstein 107867
Formula Weight 125.19
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-5-vinylthiazole
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 217-160-4
RTECSNumber XJ5104000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only