missing translation for 'onlineSavingsMsg'
Learn More

4'-Methyl-4-pentylbiphenyl 98.0+%, TCI America™

Numéro de catalogue. M28855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M28855G 5 g
M28851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M28855G Fournisseur TCI America Code fournisseur M28855G
Il en reste null

Identifiants chimiques

CAS 64835-63-8
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD11053476
InChI Key ZGBOHJUSTPZQPL-UHFFFAOYSA-N
Synonym 4-Amyl-4′-methylbiphenyl
PubChem CID 103038
IUPAC Name 4-methyl-4'-pentyl-1,1'-biphenyl
SMILES CCCCCC1=CC=C(C=C1)C1=CC=C(C)C=C1

Spécifications

Melting Point 49°C
Color White
Quantity 5 g
Formula Weight 238.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Methyl-4-pentylbiphenyl