missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-penten-2-one (contains 4-Methyl-4-penten-2-one) 85.0+%, TCI America™

Catalog No. M1340500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1340500ML 500 mL
M134025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1340500ML Supplier TCI America Supplier No. M1340500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 141-79-7
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00008900
InChI Key SHOJXDKTYKFBRD-UHFFFAOYSA-N
Synonym mesityl oxide, 4-methyl-3-penten-2-one, 3-penten-2-one, 4-methyl, isopropylideneacetone, isobutenyl methyl ketone, methyl isobutenyl ketone, mesityloxid, mesityloxyde, 3-isohexen-2-one, isopropylidene acetone
PubChem CID 8858
IUPAC Name 4-methylpent-3-en-2-one
SMILES CC(=CC(=O)C)C

Specifications

Quantity 500 mL
UN Number 1229
Formula Weight 98.15
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methyl-3-penten-2-one (contains 4-Methyl-4-penten-2-one)
EINECSNumber (2)-0566
RTECSNumber SB4200000
TSCA Yes