missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitrophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M11271G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11271G Supplier TCI America Supplier No. M11271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Methyl-3-nitrophenylboronic Acid (contains varying amounts of Anhydride)
CAS 80500-27-2
Molecular Formula C7H8BNO4
Quantity 1 g
MDL Number MFCD00191550
Synonym 4-methyl-3-nitrophenyl boronic acid, 4-methyl-3-nitrobenzeneboronic acid, 3-nitro-p-tolylboronic acid, 4-methyl-3-nitrophenyl boranediol, 4-methyl-3-nitro-phenyl boronic acid, 3-nitro-4-methylphenylboronic acid, 2-nitrotoluene-4-boronic acid, boronic acid, 4-methyl-3-nitrophenyl, 3-nitro-4-methylbenzeneboronic acid
InChI Key OASVXBRTNVFKFS-UHFFFAOYSA-N
SMILES B(C1=CC(=C(C=C1)C)[N+](=O)[O-])(O)O
IUPAC Name (4-methyl-3-nitrophenyl)boronic acid
Molecular Weight (g/mol) 180.954
PubChem CID 2773515
Formula Weight 180.95
Physical Form Crystalline Powder
Show More Show Less