missing translation for 'onlineSavingsMsg'
Learn More

4-Méthyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbaldehyde, 95%, Thermo Scientific™

Numéro de catalogue. CC62004DA
Modifier l'affichage
Click to view available options
Quantité:
1 g
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC62004DA 1 g Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC62004DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC62004DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 921938-80-9
Formule moléculaire C9H10N2O2
Poids moléculaire (g/mol) 178.191
Numéro MDL MFCD09817506
Clé InChI CVHLHSUUCRUZHB-UHFFFAOYSA-N
Synonyme 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carbaldehyde, pubchem16127, 3,4-dihydro-7-formyl-4-methyl-2h-pyrido 3,2-b 1,4 oxazine, 4-methyl-2h,3h,4h-pyrido 3,2-b 1,4 oxazine-7-carbaldehyde, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carbaldehyde, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazine-7-carbaldehyde, 4-methyl-2,3-dihydropyrido 3,2-b 1,4 oxazine-7-carboxaldehyde, 2h-pyrido 3,2-b-1,4-oxazine-7-carboxaldehyde,3,4-dihydro-4-methyl, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-carboxaldehyde
PubChem CID 24229647
Nom de l’IUPAC 4-méthyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carbaldehyde
SOURIRES CN1CCOC2=C1N=CC(=C2)C=O

Spécifications

Nom ou substance chimique 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carbaldehyde
CAS % minimum 94.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 178.19
Pourcentage de pureté 95%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.