missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-oxazole-5-carboxylic acid, 97%, Thermo Scientific™

Numéro de catalogue. CC18101CB
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC18101CB 250mg Amber Glass Bottle
CC18101DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC18101CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC18101CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 91137-55-2
Formule moléculaire C11H9NO3
Poids moléculaire (g/mol) 203.197
Numéro MDL MFCD01566859
Clé InChI HRFYZRHGBICKAG-UHFFFAOYSA-N
Synonyme 4-methyl-2-phenyloxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl-2-phenyl, 5-oxazolecarboxylic acid, 4-methyl-2-phenyl
PubChem CID 4176428
Nom de l’IUPAC 4-methyl-2-phenyl-1,3-oxazole-5-carboxylic acid
SOURIRES CC1=C(OC(=N1)C2=CC=CC=C2)C(=O)O

Spécifications

Nom ou substance chimique 4-Methyl-2-phenyl-1, 3-oxazole-5-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 250mg
Poids de formule 203.2
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.