missing translation for 'onlineSavingsMsg'
Learn More

4-Méthyl-2-pentyne 96,0+%, TCI America™

Catalog No. M02705ML
Change view
Click to view available options
Quantité:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
M02705ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M02705ML Supplier TCI America Supplier No. M02705ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 21020-27-9
Formule moléculaire C6H10
Poids moléculaire (g/mol) 82.15
Numéro MDL MFCD00041615
Clé InChI SLMFWJQZLPEDDU-UHFFFAOYSA-N
Synonyme 4-methyl-2-pentyne, 2-pentyne, 4-methyl, isopropylmethylacetylene, acmc-1co71
PubChem CID 140789
Nom de l’IUPAC 4-méthylpent-2-yne
SOURIRES CC#CC(C)C

Specifications

Nom ou substance chimique 4-Methyl-2-pentyne
Couleur Incolore
Point d’ébullition 72 °C
Quantité 5 mL
Numéro UN 3295
Poids de formule 82.15
Pourcentage de pureté ≥96,0% (GC)
Forme physique Liquide