missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS, 98.5+%

Catalog No. AA33346K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA33346K2 1 L
AA33346AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA33346K2 Supplier Thermo Scientific Chemicals Supplier No. 033346K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-10-1 | C6H12O | 100.161 g/mol

4-methyl-2-pentanone is used as a solvent for nitrocellulose, gums, resins, varnishes, paints, lacquers, nitrocellulose, certain polymers and as a denaturing agent. It acts as a precursor for the preparation of N-(1,3-dimethylbutyl)-N'-phenyl-p-phenylene diamine (6PPD), which finds application as an antiozonant in tires. Its aqueous solution serves as a developer for PMMA electron beam lithography resist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.801 g/mL
Boiling Point 117°C to 118°C
Flash Point 13°C (55°F)
Odor Mild
Refractive Index 1.396
Quantity 1 L
UN Number UN1245
Beilstein 605399
Merck Index 14,5207
Solubility Information Miscible with ethanol,ether,acetone,benzene and chloroform. Slightly miscible with water.
Formula Weight 100.16
Percent Purity ≥98.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material 4-Methyl-2-pentanone
Show More Show Less
Hazard Category H225-H319-H332-H335-H351
Hazard Statement GHS H Statement
H225-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280i-P281-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL ISOBUTYL KETONE
EINECSNumber 203-550-1
RTECSNumber SA9275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only