missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-pentyne, 97%

Catalog No. AAL0553703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0553703 1 g
AAL0553706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0553703 Supplier Thermo Scientific Chemicals Supplier No. L0553703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7154-75-8 | C6H10 | 82.15 g/mol

4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1-pentyne, is used as a polymer matrix together with synthesized NH2-MIL 53 metal organic framework (MOF) as a filler were used to fabricate a mixed matrix membrane (MMM). It is also used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

Solubility
Insoluble in water. Soluble in common organic solvents of low polarity.

Notes
Stable under normal temperatures and pressures. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 7154-75-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00039857
InChI Key OXRWICUICBZVAE-UHFFFAOYSA-N
Synonym 4-methyl-1-pentyne, 1-pentyne, 4-methyl, isobutylacetylene, isobutylethyne, 4-methylpentine, 4-methylpentyne, 4-methyl pentyne, isobutyl acetylene, 4-methyl-pentyne, 4-methyl-pent-1-yne
PubChem CID 138948
IUPAC Name 4-methylpent-1-yne
SMILES CC(C)CC#C

Specifications

Density 0.698
Boiling Point 61°C to 62°C
Flash Point −24°C (−11°F)
Refractive Index 1.393
Quantity 1 g
UN Number UN3295
Beilstein 1731077
Solubility Information Insoluble in water. Soluble in common organic solvents of low polarity.
Formula Weight 82.15
Percent Purity 97%
Chemical Name or Material 4-Methyl-1-pentyne
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only