missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthaleneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M24571G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24571G Supplier TCI America Supplier No. M24571G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103986-53-4
Molecular Formula C11H11BO2
Molecular Weight (g/mol) 186.02
MDL Number MFCD01632204
InChI Key JHVQEUGNYSVSDH-UHFFFAOYSA-N
PubChem CID 2773511
IUPAC Name (4-methylnaphthalen-1-yl)boronic acid
SMILES CC1=CC=C(B(O)O)C2=CC=CC=C12

Specifications

Melting Point 207°C
Color Yellow
Quantity 1 g
Synonym 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid, 1-borono-4-methylnaphthalene, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, boronic acid, 4-methyl-1-naphthalenyl
Formula Weight 186.02
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1-naphthaleneboronic Acid (contains varying amounts of Anhydride)