missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2-cyclohexene oxide, cis + trans, 97%

Catalog No. AAH2910014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2910014 25 g
AAH2910003 1 g
AAH2910006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2910014 Supplier Thermo Scientific Chemicals Supplier No. H2910014

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36099-51-1 | C7H12O | 112.172 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36099-51-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD09742280
InChI Key ULPDSNLBZMHGPI-UHFFFAOYSA-N
Synonym 3-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane, 3-methyl, 4-methyl-1,2-cyclohexene oxide, 4-methyl-1,2-cyclohexene oxide, cis + trans, 4-methyl-7-oxabicyclo 4.1.0 heptane, 7-oxabicyclo 4.1.0 heptane,3-methyl, 3-methyl-7-oxabicyclo 4.1.0 heptane #, 4-methyl-1,2-cyclohexene oxide, cis+trans
PubChem CID 535184
IUPAC Name 4-methyl-7-oxabicyclo[4.1.0]heptane
SMILES CC1CCC2C(C1)O2

Specifications

Refractive Index 1.447
Quantity 25 g
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 4-Methyl-1,2-cyclohexene oxide, Cis + Trans
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only