missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxytrityl Chloride 98.0+%, TCI America™

Catalog No. M079025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M079025G 25 g
M0790250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M079025G Supplier TCI America Supplier No. M079025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14470-28-1
Molecular Formula C20H17ClO
Molecular Weight (g/mol) 308.805
MDL Number MFCD00000814
InChI Key OBOHMJWDFPBPKD-UHFFFAOYSA-N
Synonym 4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
PubChem CID 84462
IUPAC Name 1-[chloro(diphenyl)methyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl

Specifications

Melting Point 124°C
Quantity 25 g
Formula Weight 308.81
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxytrityl Chloride [Hydroxyl Protecting Agent]
TSCA No
Recommended Storage Refrigerator