missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxythiophenol, 97%

Catalog No. AAA1600922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1600922 100 g
AAA1600906 5 g
AAA1600914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1600922 Supplier Thermo Scientific Chemicals Supplier No. A1600922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 696-63-9 | C7H8OS | 140.2 g/mol

4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM?s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxythiophenol is used in preparing samples for self-assembled monolayers (SAM′s) characterization and to study SAM’s effect on both n- and p-channel organic thin film transistors. It is also used in cesium fluoride-Celite catalyzed preparation of thioethers and thioesters.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.

Chemical Identifiers

CAS 696-63-9
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD00004849
InChI Key NIFAOMSJMGEFTQ-UHFFFAOYSA-N
Synonym 4-methoxythiophenol, p-methoxybenzenethiol, benzenethiol, 4-methoxy, 4-methoxybenzene-1-thiol, benzenethiol, p-methoxy, 4-mercaptoanisole, 4-methoxy thiophenol, 4-methoxy-benzenethiol, para-methoxybenzenethiol, 4-mercaptoanisol
PubChem CID 12765
IUPAC Name 4-methoxybenzenethiol
SMILES COC1=CC=C(C=C1)S

Specifications

Density 1.14
Boiling Point 100°C to 103°C (13 mmHg)
Flash Point 96°C (204°F)
Odor Unpleasant
Refractive Index 1.5831
Quantity 100 g
Beilstein 1446910
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 140.21
Percent Purity 97%
Chemical Name or Material 4-Methoxythiophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 211-799-2
RTECSNumber DC1800000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only