missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[(4-Methoxyphenyl)ethynyl]trimethylsilane 97.0+%, TCI America™
Supplier: TCI America M27851G
Specifications
| [(4-Methoxyphenyl)ethynyl]trimethylsilane | |
| Yellow | |
| C12H16OSi | |
| 1 g | |
| COC1=CC=C(C=C1)C#C[Si](C)(C)C | |
| 204.34 | |
| 204.34 | |
| Liquid |
| 3989-14-8 | |
| 96°C | |
| MFCD04039887 | |
| GODVAYLZXZQBFP-UHFFFAOYSA-N | |
| [2-(4-methoxyphenyl)ethynyl]trimethylsilane | |
| 3497861 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 3989-14-8 | |
| 204.34 | |
| GODVAYLZXZQBFP-UHFFFAOYSA-N | |
| [2-(4-methoxyphenyl)ethynyl]trimethylsilane |
| C12H16OSi | |
| MFCD04039887 | |
| 3497861 | |
| COC1=CC=C(C=C1)C#C[Si](C)(C)C |
Safety and Handling
TSCA : No