accessibility menu, dialog, popup

UserName

H3COSiCH3H3CCH3

CAS RN 3989-14-8

IUPAC Name: [2-(4-methoxyphenyl)ethynyl]trimethylsilane
Synonyms: [2-(4-methoxyphenyl)ethynyl]trimethylsilane
Molecular Weight (g/mol): 204.34
Molecular Formula: C12H16OSi
InChi Key: GODVAYLZXZQBFP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C#C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results