missing translation for 'onlineSavingsMsg'
Learn More

[(4-Methoxyphenyl)ethynyl]trimethylsilane 97.0+%, TCI America™

Catalog No. p-7133425
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M27851G Supplier TCI America Supplier No. M27851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3989-14-8
Molecular Formula C12H16OSi
Molecular Weight (g/mol) 204.34
MDL Number MFCD04039887
InChI Key GODVAYLZXZQBFP-UHFFFAOYSA-N
PubChem CID 3497861
IUPAC Name [2-(4-methoxyphenyl)ethynyl]trimethylsilane
SMILES COC1=CC=C(C=C1)C#C[Si](C)(C)C

Specifications

Color Yellow
Boiling Point 96°C
Quantity 1 g
Formula Weight 204.34
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material [(4-Methoxyphenyl)ethynyl]trimethylsilane