missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl chloroformate, 98%

Catalog No. p-3758326
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314570050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314570050 Supplier Thermo Scientific Chemicals Supplier No. 314570050
Only null left

CAS: 7693-41-6 | C8H7ClO3 | 186.591 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 7693-41-6
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00013258
InChI Key CCFSGQKTSBIIHG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl chloroformate, carbonochloridic acid, 4-methoxyphenyl ester, 4-methoxyphenylchloroformate, chloroformic acid 4-methoxyphenyl ester, acmc-1bfyr, p-methoxyphenyl chloroformate, 4-methoxy-phenyl chloroformate, 4-methoxy phenyl chloroformate, 4-methoxyphenyl carbonochloridate
PubChem CID 82128
IUPAC Name (4-methoxyphenyl) carbonochloridate
SMILES COC1=CC=C(C=C1)OC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.25g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5220 to 1.5260 (20°C, 589nm)
Linear Formula ClCOC6H4OCH3
Quantity 5 g
UN Number 3277
Formula Weight 186.59
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only