missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphényl 4-O-Acétyl-3,6-di-O-benzyl-2-désoxy-2-phthalimido-bêta-D-glucopyranoside 98,0+%, TCI America™

Numéro de catalogue. M18341G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M18341G 1 g
M18345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M18341G Fournisseur TCI America Code fournisseur M18341G

Identifiants chimiques

CAS 140615-77-6
Molecular Formula C37H35NO9
Molecular Weight (g/mol) 637.685
InChI Key QZTMIUALKXAHCQ-RHPHZSOCSA-N
Synonym GlcNPhth[36Bn,4Ac]-beta-MP
PubChem CID 133554185
IUPAC Name [(2R,3R,4S,5R,6R)-5-(1,3-dioxoisoindol-2-yl)-6-(4-méthoxyphénoxy)-4-phénylméthoxy-2-(phénylméthoxyméthoxy)oxan-3-yl] acétate
SMILES CC(=O)OC1C(OC(C(C1OCC2=CC=CC=C2)N3C(=O)C4=CC=CC=C4C3=O)OC5=CC=C(C=C5)OC)COCC6=CC=CC=C6

Spécifications

Color Blanc
Quantity 1 g
Formula Weight 637.69
Percent Purity ≥98.0% (HPLC)
Physical Form Poudre cristalline
Chemical Name or Material 4-Methoxyphenyl 4-O-Acetyl-3,6-di-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside
TSCA No
Recommended Storage Réfrigérateur