missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3-O-Benzyl-beta-D-galactopyranoside 98.0+%, TCI America™

Catalog No. p-7132826
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17251G 1 g
M17255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17251G Supplier TCI America Supplier No. M17251G

Chemical Identifiers

CAS 383905-60-0
Molecular Formula C20H24O7
Molecular Weight (g/mol) 376.405
InChI Key CZPFTCBIXZWFIZ-UJMXGEILSA-N
PubChem CID 77620864
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-methoxyphenoxy)-4-phenylmethoxyoxane-3,5-diol
SMILES COC1=CC=C(C=C1)OC2C(C(C(C(O2)CO)O)OCC3=CC=CC=C3)O

Specifications

Melting Point 154°C
Color White
Quantity 1 g
Formula Weight 376.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3-O-Benzyl-beta-D-galactopyranoside
TSCA No
Recommended Storage Refrigerator