missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl 3-O-Allyl-6-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside 98.0+%, TCI America™

Catalog No. M16041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16041G 1 g
M16045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16041G Supplier TCI America Supplier No. M16041G

Chemical Identifiers

CAS 1820583-64-9
Molecular Formula C31H31NO8
Molecular Weight (g/mol) 545.59
MDL Number MFCD16295191
InChI Key UGQLXVORDHWCEW-UHFFFAOYNA-N
PubChem CID 91659133
IUPAC Name 2-{6-[(benzyloxy)methyl]-5-hydroxy-2-(4-methoxyphenoxy)-4-(prop-2-en-1-yloxy)oxan-3-yl}-2,3-dihydro-1H-isoindole-1,3-dione
SMILES COC1=CC=C(OC2OC(COCC3=CC=CC=C3)C(O)C(OCC=C)C2N2C(=O)C3=CC=CC=C3C2=O)C=C1

Specifications

Color White
Quantity 1 g
Formula Weight 545.59
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenyl 3-O-Allyl-6-O-benzyl-2-deoxy-2-phthalimido-beta-D-glucopyranoside